3-Iod-4-methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS1864G1 AS1864G5
ACTIAS1864G1EA 0 CHF
ACTIAS1864G1 ACTIAS1864G5
3-Iod-4-methyl-1H-indazol
3-Iodo-4-methyl-1H-indazole
Formel: C₈H₇IN₂
MW: 258.0591 g/mol
MDL Number: MFCD07781537
CAS-Nummer: 885522-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR