3-Amino-5-fluor-1-methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Fluoro-1-methyl-1H-indazol-3-amine

AS50725G1 AS50725G5
ACTIAS50725G1EA 0 CHF
ACTIAS50725G1 ACTIAS50725G5
3-Amino-5-fluor-1-methyl-1H-indazol
3-Amino-5-fluoro-1-methyl-1H-indazole
Formel: C₈H₈FN₃
MW: 165,1700 g/mol
MDL Number: MFCD09054760
CAS-Nummer: 171809-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR