3-Amino-4-fluor-1-methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS50723G1 AS50723G5
ACTIAS50723G1EA 0 CHF
ACTIAS50723G1 ACTIAS50723G5
3-Amino-4-fluor-1-methyl-1H-indazol
3-Amino-4-fluoro-1-methyl-1H-indazole
Formel: C₈H₈FN₃
MDL Number: MFCD11109397
CAS-Nummer: 162502-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR