5-Amino-3-brom-1-methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-1-methyl-1H-indazol-5-amine

AS50617G1 AS50617G5
ACTIAS50617G1EA 0 CHF
ACTIAS50617G1 ACTIAS50617G5
5-Amino-3-brom-1-methyl-1H-indazol
5-Amino-3-bromo-1-methyl-1H-indazole
Formel: C₈H₈BrN₃
MW: 226,0700 g/mol
MDL Number: MFCD11100026
CAS-Nummer: 1092351-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR