7-Brom-2-methyl-2H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS50377G1 AS50377G5
ACTIAS50377G1EA 0 CHF
ACTIAS50377G1 ACTIAS50377G5
7-Brom-2-methyl-2H-indazol
7-Bromo-2-methyl-2H-indazole
Formel: C₈H₇BrN₂
MDL Number: MFCD09870049
CAS-Nummer: 701910-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR