7-Brom-1-methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 7-Bromo-1-methylindazole

AS1910G1 AS1910G5
ACTIAS1910G1EA 0 CHF
ACTIAS1910G1 ACTIAS1910G5
7-Brom-1-methyl-1H-indazol
7-Bromo-1-methyl-1H-indazole
Formel: C₈H₇BrN₂
MW: 211,0600 g/mol
MDL Number: MFCD09878570
CAS-Nummer: 1000576-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR