6-Brom-2-methyl-2H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS50376G1 AS50376G5
ACTIAS50376G1EA 0 CHF
ACTIAS50376G1 ACTIAS50376G5
6-Brom-2-methyl-2H-indazol
6-Bromo-2-methyl-2H-indazole
Formel: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD09870048
CAS-Nummer: 590417-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR