5-Brom-6-methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS1890G1 AS1890G5
ACTIAS1890G1EA 0 CHF
ACTIAS1890G1 ACTIAS1890G5
5-Brom-6-methyl-1H-indazol
5-Bromo-6-methyl-1H-indazole
Formel: C₈H₇BrN₂
MW: 211,0600 g/mol
MDL Number: MFCD09026991
CAS-Nummer: 885223-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR