5-Brom-4-methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS51013G1 AS51013G5
ACTIAS51013G1EA 0 CHF
ACTIAS51013G1 ACTIAS51013G5
5-Brom-4-methyl-1H-indazol
5-Bromo-4-methyl-1H-indazole
Formel: C₈H₇BrN₂
MDL Number: MFCD11007842
CAS-Nummer: 1082041-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR