5-Brom-3-methyl-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS1685G1 AS1685G5
ACTIAS1685G1EA 0 CHF
ACTIAS1685G1 ACTIAS1685G5
5-Brom-3-methyl-1H-indazol
5-Bromo-3-methyl-1H-indazole
Formel: C₈H₇BrN₂
MW: 211,0600 g/mol
MDL Number: MFCD05664003
CAS-Nummer: 552331-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR