5-Brom-2-methyl-2H-indazol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Methyl-5-bromo-2H-indazole
AS50375G1
AS50375G5
ACTIAS50375G1EA
0
CHF
ACTIAS50375G1
ACTIAS50375G5
5-Brom-2-methyl-2H-indazol
5-Bromo-2-methyl-2H-indazole
Formel:
C₈H₇BrN₂ MW: 211,0600 g/mol |
MDL Number:
MFCD09870047 CAS-Nummer: 465529-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...