4-Brom-2-methyl-2H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS50374G1 AS50374G5
ACTIAS50374G1EA 0 CHF
ACTIAS50374G1 ACTIAS50374G5
4-Brom-2-methyl-2H-indazol
4-Bromo-2-methyl-2H-indazole
Formel: C₈H₇BrN₂
MW: 211.0586 g/mol
MDL Number: MFCD09870046
CAS-Nummer: 590417-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR