5-Methyl-1H-indazol-4-boronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (5-Methyl-1H-indazol-4-yl)boronic acid

AS1897G1 AS1897G5
ACTIAS1897G1EA 0 CHF
ACTIAS1897G1 ACTIAS1897G5
5-Methyl-1H-indazol-4-boronsäure
5-Methyl-1H-indazole-4-boronic acid
Formel: C₈H₉BN₂O₂
MDL Number: MFCD10001490
CAS-Nummer: 1245816-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR