4-Methyl-1H-indazol-5-boronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Methyl-1H-indazol-5-yl)boronic acid

AS51083G1 AS51083G5
ACTIAS51083G1EA 0 CHF
ACTIAS51083G1 ACTIAS51083G5
4-Methyl-1H-indazol-5-boronsäure
4-Methyl-1H-indazole-5-boronic acid
Formel: C₈H₉BN₂O₂
MW: 175,9800 g/mol
MDL Number: MFCD12028644
CAS-Nummer: 1245816-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR