2-Methyl-2H-indazol-5-boronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-Methyl-2H-indazol-5-yl)boronic acid

AS12293G1 AS12293G5
ACTIAS12293G1EA 0 CHF
ACTIAS12293G1 ACTIAS12293G5
2-Methyl-2H-indazol-5-boronsäure
2-Methyl-2H-indazole-5-boronic acid
Formel: C₈H₉BN₂O₂
MDL Number: MFCD09870056
CAS-Nummer: 952319-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR