2-Methyl-2H-indazol-6-boronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

AS50789G1 AS50789G5
ACTIAS50789G1EA 0 CHF
ACTIAS50789G1 ACTIAS50789G5
2-Methyl-2H-indazol-6-boronsäurepinakolester
2-Methyl-2H-indazol-6-boronsäurepinakolester
Formel: C₁₄H₁₉BN₂O₂
MW: 258.1239 g/mol
MDL Number: MFCD11109435
CAS-Nummer: 1204580-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR