2-Methyl-2H-indazol-4-boronsäurepinakolester
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole
AS50791G1
AS50791G5
ACTIAS50791G1EA
0
CHF
ACTIAS50791G1
ACTIAS50791G5
2-Methyl-2H-indazol-4-boronsäurepinakolester
2-Methyl-2H-indazol-4-boronsäurepinakolester
Formel:
C₁₄H₁₉BN₂O₂ MW: 258,1239 g/mol |
MDL Number:
MFCD11109437 CAS-Nummer: 885698-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...