6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS22956G25 AS22956G5
ACTIAS22956G25EA 0 CHF
ACTIAS22956G25 ACTIAS22956G5
6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure
6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure
Formel: C₅H₆N₂O₃
MW: 142.12 g/mol
MDL Number: MFCD00052030
CAS-Nummer: 27372-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR