(1H-Pyrazol-1-yl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Pyrazoleacetic Acid

AS35455G1 AS35455G5
ACTIAS35455G1EA 0 CHF
ACTIAS35455G1 ACTIAS35455G5
(1H-Pyrazol-1-yl)essigsäure
(1H-Pyrazol-1-yl)essigsäure
Formel: C₅H₆N₂O₂
MW: 126.1133 g/mol
MDL Number: MFCD00228774
CAS-Nummer: 16034-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR