Imidazo[1,2-a]pyridin-2-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS1103G1 AS1103G5
ACTIAS1103G1EA 0 CHF
ACTIAS1103G1 ACTIAS1103G5
Imidazo[1,2-a]pyridin-2-carbonsäure
Imidazo[1,2-a]pyridin-2-carbonsäure
Formel: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD03419462
CAS-Nummer: 64951-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR