Ethyl-6-aminoimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 6-aminoimidazo[1,2-a]pyridine-2-carboxylate

AS30222G1 AS30222G5
ACTIAS30222G1EA 0 CHF
ACTIAS30222G1 ACTIAS30222G5
Ethyl-6-aminoimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-6-aminoimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₀H₁₁N₃O₂
MW: 205,2132 g/mol
MDL Number: MFCD05864804
CAS-Nummer: 158980-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR