Methylimidazo[1,2-a]pyridin-6-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS21968G25 AS21968G5
ACTIAS21968G25EA 0 CHF
ACTIAS21968G25 ACTIAS21968G5
Methylimidazo[1,2-a]pyridin-6-carboxylat
Methylimidazo[1,2-a]pyridin-6-carboxylat
Formel: C₉H₈N₂O₂
MDL Number: MFCD06200984
CAS-Nummer: 136117-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR