Imidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS51417G1 AS51417G5
ACTIAS51417G1EA 0 CHF
ACTIAS51417G1 ACTIAS51417G5
Imidazo[1,2-a]pyridin-3-carbaldehyd
Imidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₈H₆N₂O
MW: 146.1500 g/mol
MDL Number: MFCD07778354
CAS-Nummer: 6188-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR