Imidazo[1,2-b]pyridazin-2-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS2609G1 AS2609G5
ACTIAS2609G1EA 268 CHF
ACTIAS2609G1 ACTIAS2609G5
Imidazo[1,2-b]pyridazin-2-carbonsäure
Imidazo[1,2-b]pyridazin-2-carbonsäure
Formel: C₇H₅N₃O₂
MW: 163,1300 g/mol
MDL Number: MFCD08235229
CAS-Nummer: 160911-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR