4-(Ethylsulfonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-(Ethylsulphonyl)phenyl)boronic acid, 4-(Ethylsulphonyl)benzeneboronic acid

AS51361G1 AS51361G5
ACTIAS51361G1EA 0 CHF
ACTIAS51361G1 ACTIAS51361G5
4-(Ethylsulfonyl)phenylboronsäure
4-(Ethylsulfonyl)phenylboronsäure
Formel: C₈H₁₁BO₄S
MW: 214.05 g/mol
MDL Number: MFCD02093076
CAS-Nummer: 352530-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR