3-(Methylsulfonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-(Methanesulphonyl)phenylboronic acid, 3-(Methylsulphonyl)benzeneboronic acid, (3-(Methylsulphonyl)phenyl)boronic acid

AS50886G1 AS50886G5
ACTIAS50886G1EA 0 CHF
ACTIAS50886G1 ACTIAS50886G5
3-(Methylsulfonyl)phenylboronsäure
3-(Methylsulfonyl)phenylboronsäure
Formel: C₇H₉BO₄S
MW: 200,02 g/mol
MDL Number: MFCD03092935
CAS-Nummer: 373384-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR