3-(Ethylsulfonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50181G1 AS50181G5
ACTIAS50181G1EA 0 CHF
ACTIAS50181G1 ACTIAS50181G5
3-(Ethylsulfonyl)phenylboronsäure
3-(Ethylsulfonyl)phenylboronsäure
Formel: C₈H₁₁BO₄S
MDL Number: MFCD03095257
CAS-Nummer: 845870-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR