Methyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS14376G1 AS14376G5
ACTIAS14376G1EA 0 CHF
ACTIAS14376G1 ACTIAS14376G5
Methyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat
Methyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat
Formel: C₉H₇BrN₂O₂
MDL Number: MFCD21364589
CAS-Nummer: 1359656-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR