Ethyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 6-bromoimidazo[1,2-a]pyridine-3-carboxylate

AS14377G1 AS14377G5
ACTIAS14377G1EA 0 CHF
ACTIAS14377G1 ACTIAS14377G5
Ethyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat
Ethyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat
Formel: C₁₀H₉BrN₂O₂
MW: 269.1000 g/mol
MDL Number: MFCD10698068
CAS-Nummer: 372198-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR