Methyl-2-bromimidazo[1,2-a]pyridin-6-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-bromoimidazo[1,2-a]pyridine-6-carboxylate

AS32084G1 AS32084G5
ACTIAS32084G1EA 0 CHF
ACTIAS32084G1 ACTIAS32084G5
Methyl-2-bromimidazo[1,2-a]pyridin-6-carboxylat
Methyl-2-bromimidazo[1,2-a]pyridin-6-carboxylat
Formel: C₉H₇BrN₂O₂
MW: 255,0700 g/mol
MDL Number: MFCD13195435
CAS-Nummer: 1042141-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR