2-(Methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(2-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Methoxycarbonylphenylboronic acid pinacol ester

AS20743G1 AS20743G5
ACTIAS20743G1EA 0 CHF
ACTIAS20743G1 ACTIAS20743G5
2-(Methoxycarbonyl)phenylboronsäurepinakolester
2-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₄
MDL Number: MFCD05663866
CAS-Nummer: 653589-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR