5-(4-Hydroxyphenyl)-1H-tetrazol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-(1H-Tetrazol-5-yl)phenol
AS23665G1
AS23665G5
ACTIAS23665G1EA
125
CHF
ACTIAS23665G1
ACTIAS23665G5
5-(4-Hydroxyphenyl)-1H-tetrazol
5-(4-Hydroxyphenyl)-1H-tetrazole
Formel:
C₇H₆N₄O MW: 162,16 g/mol |
MDL Number:
MFCD05662709 CAS-Nummer: 51517-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...