5-(2-Hydroxyphenyl)-1H-tetrazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(1H-Tetrazol-5-yl)phenol

AS23666G1 AS23666G5
ACTIAS23666G1EA 111 CHF
ACTIAS23666G1 ACTIAS23666G5
5-(2-Hydroxyphenyl)-1H-tetrazol
5-(2-Hydroxyphenyl)-1H-tetrazole
Formel: C₇H₆N₄O
MW: 162,16 g/mol
MDL Number: MFCD09037787
CAS-Nummer: 51449-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR