N-Fmoc-L-norleucin

Lieferant: BACHEM BIOCHEMICA

BACE4018558.0001EA 0 CHF
BACE4018558.0001 BACE4018558.0005
N-Fmoc-L-norleucin
N-Fmoc-L-norleucin
Formel: C₂₁H₂₃NO₄
MW: 353,4 g/mol
MDL Number: MFCD00037537
CAS-Nummer: 77284-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR