2-Hydroxy-5-(trifluormethyl)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS35600G1 AS35600G5
ACTIAS35600G1EA 0 CHF
ACTIAS35600G1 ACTIAS35600G5
2-Hydroxy-5-(trifluormethyl)benzonitril
2-Hydroxy-5-(trifluormethyl)benzonitril
Formel: C₈H₄F₃NO
MDL Number: MFCD11213679
CAS-Nummer: 142167-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR