3-Hydroxy-4-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS21831G1 AS21831G25 AS21831G5
ACTIAS21831G1EA 0 CHF
ACTIAS21831G1 ACTIAS21831G25 ACTIAS21831G5
3-Hydroxy-4-methoxybenzonitril
3-Hydroxy-4-methoxybenzonitril
Formel: C₈H₇NO₂
MDL Number: MFCD02258854
CAS-Nummer: 52805-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR