2-Hydroxy-4-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS31426G1 AS31426G5
ACTIAS31426G1EA 0 CHF
ACTIAS31426G1 ACTIAS31426G5
2-Hydroxy-4-methoxybenzonitril
2-Hydroxy-4-methoxybenzonitril
Formel: C₈H₇NO₂
MDL Number: MFCD08567959
CAS-Nummer: 39835-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR