1-Methyl-5-indolboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Methylindole-5-boronic acid, N-Methylindole-5-boronic acid

AS50877G1 AS50877G5
ACTIAS50877G1EA 0 CHF
ACTIAS50877G1 ACTIAS50877G5
1-Methyl-5-indolboronsäure
1-Methyl-5-indoleboronic acid
Formel: C₉H₁₀BNO₂
MW: 174.9900 g/mol
MDL Number: MFCD03412070
CAS-Nummer: 192182-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR