2-Brom-3-nitrotoluol

Lieferant: ACTIVATE SCIENTIFIC

AS1479G100 AS1479G5
ACTIAS1479G100EA 0 CHF
ACTIAS1479G100 ACTIAS1479G5
2-Brom-3-nitrotoluol
2-Brom-3-nitrotoluol
Formel: C₇H₆BrNO₂
MW: 216,0300 g/mol
MDL Number: MFCD00134555
CAS-Nummer: 41085-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR