2-Brom-3-nitroanisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-1-methoxy-3-nitrobenzene

AS40171G1 AS40171G5
ACTIAS40171G1EA 0 CHF
ACTIAS40171G1 ACTIAS40171G5
2-Brom-3-nitroanisol
2-Brom-3-nitroanisol
Formel: C₇H₆BrNO₃
MW: 232,0300 g/mol
MDL Number: MFCD07779272
CAS-Nummer: 67853-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR