2'-(Trifluormethyl)biphenyl-4-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

AS30291G1 AS30291G5
ACTIAS30291G1EA 0 CHF
ACTIAS30291G1 ACTIAS30291G5
2'-(Trifluormethyl)biphenyl-4-carbonsäure
2'-(Trifluormethyl)biphenyl-4-carbonsäure
Formel: C₁₄H₉F₃O₂
MW: 266,22 g/mol
MDL Number: MFCD03424605
CAS-Nummer: 198205-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR