2'-(Trifluormethyl)biphenyl-3-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid

AS30292G1 AS30292G5
ACTIAS30292G1EA 0 CHF
ACTIAS30292G1 ACTIAS30292G5
2'-(Trifluormethyl)biphenyl-3-carbonsäure
2'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel: C₁₄H₉F₃O₂
MW: 266,22 g/mol
MDL Number: MFCD03424602
CAS-Nummer: 168618-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR