2'-(Trifluormethyl)biphenyl-4-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30288G1 AS30288G5
ACTIAS30288G1EA 0 CHF
ACTIAS30288G1 ACTIAS30288G5
2'-(Trifluormethyl)biphenyl-4-carbaldehyd
2'-(Trifluormethyl)biphenyl-4-carbaldehyd
Formel: C₁₄H₉F₃O
MDL Number: MFCD01631916
CAS-Nummer: 198205-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR