2'-(Trifluormethyl)biphenyl-3-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carbaldehyde

AS30294G1 AS30294G5
ACTIAS30294G1EA 0 CHF
ACTIAS30294G1 ACTIAS30294G5
2'-(Trifluormethyl)biphenyl-3-carbaldehyd
2'-(Trifluormethyl)biphenyl-3-carbaldehyd
Formel: C₁₄H₉F₃O
MW: 250,2159 g/mol
MDL Number: MFCD03424642
CAS-Nummer: 675596-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR