4-Fluor-3-iodbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS33810G1 AS33810G5
ACTIAS33810G1EA 0 CHF
ACTIAS33810G1 ACTIAS33810G5
4-Fluor-3-iodbenzoesäure
4-Fluoro-3-iodobenzoic acid
Formel: C₇H₄FIO₂
MW: 266.01 g/mol
MDL Number: MFCD03788556
CAS-Nummer: 403-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR