5-Fluor-2-iodbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS31310G1 AS31310G5
ACTIAS31310G1EA 0 CHF
ACTIAS31310G1 ACTIAS31310G5
5-Fluor-2-iodbenzoesäure
5-Fluor-2-iodbenzoesäure
Formel: C₇H₄FIO₂
MW: 266,01 g/mol
MDL Number: MFCD00837316
CAS-Nummer: 52548-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR