3-Fluor-4-iodbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS33898G1 AS33898G5
ACTIAS33898G1EA 0 CHF
ACTIAS33898G1 ACTIAS33898G5
3-Fluor-4-iodbenzoesäure
3-Fluoro-4-iodobenzoic acid
Formel: C₇H₄FIO₂
MW: 266,0100 g/mol
MDL Number: MFCD07774250
CAS-Nummer: 825-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR