3-Fluor-2-iodbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS33802G1 AS33802G5
ACTIAS33802G1EA 0 CHF
ACTIAS33802G1 ACTIAS33802G5
3-Fluor-2-iodbenzoesäure
3-Fluoro-2-iodobenzoic acid
Formel: C₇H₄FIO₂
MW: 266,0100 g/mol
MDL Number: MFCD07774249
CAS-Nummer: 387-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR