2-Brom-5-chlorbenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-Bromo-5-chlorophenyl)methanol

AS35537G1 AS35537G5
ACTIAS35537G1EA 0 CHF
ACTIAS35537G1 ACTIAS35537G5
2-Brom-5-chlorbenzylalkohol
2-Brom-5-chlorbenzylalkohol
Formel: C₇H₆BrClO
MW: 221.4800 g/mol
MDL Number: MFCD09261147
CAS-Nummer: 60666-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR