Ethyl-2,4-dimethylbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 2,4-dimethylbenzoate

AS20795G25 AS20795G5
ACTIAS20795G25EA 261 CHF
ACTIAS20795G25 ACTIAS20795G5
Ethyl-2,4-dimethylbenzoat
Ethyl-2,4-dimethylbenzoat
Formel: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00015438
CAS-Nummer: 33499-42-2
EINECS: 251-544-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR